Learn More
SeaKem™ HE Agarose, Lonza™
Click to view available options
Quantity:
25 g
125 g
Unit Size:
Each
Description
- For immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CEIP), and serum protein electrophoresis
- High EEO (0.23 –0.26)
- Standard gelling temperature agarose
Specifications
Specifications
| Chemical Name or Material | SeaKem HE Agarose |
| Quantity | 25 g |