Learn More
Lonza™ Latitude™ Precast Agarose Midigels
Product Code. 11597875
Click to view available options
Separation Range:
100 to ≥2000 bp
300 to ≥8000 bp
Wells:
2 x 20-well
20-well
Unit Size:
Each
Pack of 8
Product Code. 11597875
Brand: Lonza™ 57231
Efficiently resolve DNA ranging from 10bp to more than 10kb. Latitude™ Precast Agarose Midigels are designed for accurate DNA fragment separation for most any gel electrophoresis application.
  • Ideal for checking restriction digests and PCR products
  • Quality and performance tested to ensure reproducible results every time
  • Prestained with ethidium bromide
  • Compatible with common midichambers, multichannel pipets and alternate-well microtiter plate spacing
Note:ethidium bromide is a powerful mutagen; gloves should be worn when opening pouches and handling gels and solutions containing ethidium bromide
TRUSTED_SUSTAINABILITY
Specifications
Buffer TBE
Color Clear
Concentration 2%
Product Type Precast Gel
Separation Range 100 to ≥2000 bp
Wells 2 x 20-well
Dnase Free Yes
Formulation 2% SeaKem™ LE Plus Agarose, 1X TBE, Prestained with 0.5μg/mL of Ethidium Bromide
For Use With (Application) Designed for accurate DNA fragment separation for most any Gel electrophoresis application
Material Agarose
Quantity 8 Gels
RNase Free Yes
Storage Requirements 18° to 26°C
Thickness (English) 0.23 in.
Thickness (Metric) 0.6 cm
Volume Per Well 10 to 12 μL