Learn More
Lonza™ GelBond™ PAG Film for Acrylamide Gels
Click to view available options
Quantity:
10/Pk.
50/Pk.
Length (English):
6.2 in.
7 in.
10.1 in.
10.2 in.
10.3 in.
Width (English):
4.8 in.
5.4 in.
6.3 in.
6.4 in.
7.8 in.
7.9 in.
13.7 in.
Unit Size:
10 sheets
50 sheets
Description
- Covalently attaches to the acrylamide monomers during the polymerization reaction
- Makes gels easier to handle
- Helps gels retain their original dimensions during staining, destaining, and drying
Specifications
Specifications
| Gel Size | 20.3 x 26 cm |
| Product Type | PAG Film for Acrylamide Gels |
| Material | Polyester |
| Quantity | 50/Pk. |
| Thickness (English) | 0.007 in. |
| Thickness (Metric) | 0.02 cm |
| Length (English) | 10.2 in. |
| Length (Metric) | 26 cm |
| Width (English) | 7.9 in. |
| Width (Metric) | 26 cm |