Learn More
Lonza™ FlashGel™ System Recovery Cassettes
Product Code. 11592106
Click to view available options
Separation Range:
10 to 1000 bp
50 to 10,000 bp
Unit Size:
Each
Pack of 9
Product Code. 11592106
Brand: Lonza™ 57022
Eliminates agarose gel preparation, band excision and purification. Lonza™ FlashGel™ - System Recovery Cassettes deliver recovery, free from inhibitors and UV-induced damage, in a simple 5-10 minute protocol.
  • Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
  • Recover samples directly, without band excision or purification
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications
Concentration 2.2%
Product Type System Recovery Cassettes
Separation Range 10 to 1000 bp
Wells 8 + 1-well
Formulation 2.2% Agarose
For Use With (Application) Subsequent re-amplification, cloning or other techniques
Kit Contents Agarose Gel, Stain and Buffer
Material Agarose
Quantity 9 Pack
Storage Requirements 18° to 26°C
Length (English) Cassette 4.5 in.
Length (English) Gel 2.7 in.
Length (Metric) Cassette 11.5 cm
Length (Metric) Gel 7 cm
Width (English) Cassette 4.2 in.
Width (Metric) Cassette 10.7 cm
Width (Metric) Gel 8.4 cm
Height (English) Cassette 0.6 in.
Height (Metric) Cassette 1.7 cm