Learn More
Lonza™ FlashGel™ Recovery Kit
Click to view available options
Quantity:
9 Pack
Unit Size:
1 set
Description
Includes
FlashGel Recovery Cassettes, 1.2% agarose (9 Pk.), 8 + 1 well double tier; FlashGel Recovery Buffer; FlashGel Loading Dye; FlashGel QuantLadder; Visualization Glasses; FlashGel Mask
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
Specifications
| Product Type | Recovery Kit |
| For Use With (Application) | Direct DNA recovery |
| Kit Contents | FlashGel™ recovery cassettes 1.2%, 8+1 double-tier well, FlashGel™ loading dye, FlashGel™ recovery Buffer, FlashGel™ quantladder, visualization Glasses and FlashGel |
| Quantity | 9 Pack |