missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ CHO (Chinese Hamster Ovary) Serum-Free Culture Media: ProCHO4
For optimized expression of protein in CHO cells
Brand: Lonza™ BE12-029Q
Additional Details : Weight : 0.64000kg
Description
- ProCHO-4 is designed for the concurrent transition of adherent CHO cells to serum-free and suspension culture; supports faster doubling times
†Store as detailed on label.
Specifications
2° to 8°C | |
Liquid | |
No Hypoxanthine, No Thymidine, No Glutamine |
CHO | |
Phenol Red, 0.1% Pluronic F-68 | |
1 L |