Learn More
Lonza™ BioWhittaker™ CHO (Chinese Hamster Ovary) Serum-Free Culture Media: ProCHO4
For optimized expression of protein in CHO cells
133.00 EUR - 220.00 EUR
Specifications
Content And Storage 2° to 8°C
Cell Line CHO
Form Liquid
Quantity 1 L
View More Specs
Products 2
Product Code Brand Quantity With Additives Without Additives Price Quantity & Availability  
Product Code Brand Quantity With Additives Without Additives Price Quantity & Availability  
11675140
View Documents Promotion Details
Lonza™
04-919Q
1 L None No Phenol Red, No 0.1% Pluronic F-68, No Glutamine, No Hypoxanthine, No Thymidine
220.00 EUR
1 L
View alternative products
Save Up to 
»
Estimated Shipment: 30-05-2024
to see stock.
11645160
View Documents Promotion Details
Lonza™
BE12-029Q
1 L Phenol Red, 0.1% Pluronic F-68 No Hypoxanthine, No Thymidine, No Glutamine This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • ProCHO-4 is designed for the concurrent transition of adherent CHO cells to serum-free and suspension culture; supports faster doubling times
†Store as detailed on label.
Specifications
Specifications
2° to 8°C
CHO
Liquid
1 L