missing translation for 'onlineSavingsMsg'
Learn More

LOM612, MedChemExpress

Product Code. 16469786
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16469786

Brand: Medchem Express HY101035

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LOM612 is a potent FOXO relocator, with an EC50 value of 1.5 μM in U2fox RELOC cells.

LOM612 is a potent FOXO relocator, with an EC50 value of 1.5 μM in U2fox RELOC cells.

Specifications

Chemical Name or Material LOM612
Physical Form Solid
CAS 2173232-79-4
Color Red
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C13H10N2O2S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 6 mg/mL (23.23 mM; Need ultrasonic)
SMILES O=C1C2=C(SN=C2N(C)C)C(C3=CC=CC=C31)=O
Molecular Weight (g/mol) 258.3
Formula Weight 258.3
Percent Purity 98.68%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.