Learn More
Lithium Solution for ICP, 100mg/L, HNO3, Fisher Chemical™
BIOPHARMA
Product Code. 12976194 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 12976194
Brand: Fisher Chemical S 130 500
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Solution Contains HNO3
Concentration 1 g/L
Quantity 500 mL