Learn More
Linsitinib, MedChemExpress
Product Code. 16460027
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460027 50 mg 50mg
16436367 5 mg 5mg
16413638 10 mg 10mg
16484716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16460027 Supplier Medchem Express Supplier No. HY10191
MedChemExpress Linsitinib (OSI-906) is a potent, selective and orally bioavailable dual inhibitor of the IGF-1 receptor and insulin receptor (IR) with IC50s of 35 and 75 nM, respectively.
Linsitinib (OSI-906) is a potent, selective and orally bioavailable dual inhibitor of the IGF-1 receptor and insulin receptor (IR) with IC50s of 35 and 75 nM, respectively.
Specifications
Chemical Name or Material Linsitinib
CAS 867160-71-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H23N5O
Quantity 50 mg
Synonym OSI-906
Solubility Information DMSO : 50 mg/mL (118.63 mM; Need ultrasonic)
SMILES O[C@@]1(C)C[C@@H](C2=NC(C3=CC=C4C=CC(C5=CC=CC=C5)=NC4=C3)=C6C(N)=NC=CN62)C1
Molecular Weight (g/mol) 421.49
Formula Weight 421.49
Percent Purity 98.64%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease