missing translation for 'onlineSavingsMsg'
Learn More

Linear Daptomycin Hydrochloride Salt, TRC

Product Code. 30009903
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009903 2.5 mg 2.5mg
30010720 25 mg 25mg
30003382 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009903 Supplier Toronto Research Chemicals Supplier No. TRCL4664752.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C72 H103 N17 O27 . x(Cl H)
Molecular Weight (g/mol) 1675.1
Synonym (αS)-N-(1-Oxodecyl)-L-tryptophyl-D-asparaginyl-L-α-aspartyl-L-threonylglycyl-L-ornithyl-L-α-aspartyl-D-alanyl-L-α-aspartylglycyl-D-seryl-(3R)-3-methyl-L-α-glutamyl-α,2-diamino-γ-oxo-benzenebutanoic Acid Hydrochloride Salt
IUPAC Name (3R,4S)-4-[[(2R)-2-[[2-[[(2S)-2-[[(2R)-2-[[(2S)-2-[[(2S)-5-amino-2-[[2-[[(2S,3R)-2-[[(2S)-2-[[(2R)-4-amino-2-[[(2S)-2-(decanoylamino)-3-(1H-indol-3-yl)propanoyl]amino]-4-oxo-butanoyl]amino]-3-carboxy-propanoyl]amino]-3-hydroxy-butanoyl]amino]acetyl]amino]pentanoyl]amino]-3-carboxy-propanoyl]amino]propanoyl]amino]-3-carboxy-propanoyl]amino]acetyl]amino]-3-hydroxy-propanoyl]amino]-5-[[(1S)-3-(2-aminophenyl)-1-carboxy-3-oxo-propyl]amino]-3-methyl-5-oxo-pentanoic acid;hydrochloride
SMILES Cl.CCCCCCCCCC(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@H](CC(=O)N)C(=O)N[C@@H](CC(=O)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](CCCN)C(=O)N[C@@H](CC(=O)O)C(=O)N[C@H](C)C(=O)N[C@@H](CC(=O)O)C(=O)NCC(=O)N[C@H](CO)C(=O)N[C@@H]([C@H](C)CC(=O)O)C(=O)N[C@@H](CC(=O)c3ccccc3N)C(=O)O

Specifications

Analyte or Component Names Linear Daptomycin Hydrochloride Salt
Shipping Condition Room Temperature
InChI Formula InChI=1S/C72H103N17O27.ClH/c1-5-6-7-8-9-10-11-22-53(94)81-44(25-38-31-76-42-20-15-13-17-39(38)42)66(109)84-45(27-52(75)93)67(110)86-48(30-59(103)104)68(111)89-61(37(4)91)70(113)78-33-54(95)80-43(21-16-23-73)64(107)85-47(29-58(101)102)65(108)79-36(3)62(105)83-46(28-57(99)100)63(106)77-32-55(96)82-50(34-90)69(112)88-60(35(2)24-56(97)98)71(114)87-49(72(115)116)26-51(92)40-18-12-14-19-41(40)74;/h12-15,17-20,31,35-37,43-50,60-61,76,90-91H,5-11,16,21-30,32-34,73-74H2,1-4H3,(H2,75,93)(H,77,106)(H,78,113)(H,79,108)(H,80,95)(H,81,94)(H,82,96)(H,83,105)(H,84,109)(H,85,107)(H,86,110)(H,87,114)(H,88,112)(H,89,111)(H,97,98)(H,99,100)(H,101,102)(H,103,104)(H,115,116);1H/t35-,36-,37-,43+,44+,45-,46+,47+,48+,49+,50-,60+,61+;/m1./s1
Industry Type Forensics
Quantity 2.5 mg
Chemical Name or Material Linear Daptomycin Hydrochloride Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.