missing translation for 'onlineSavingsMsg'
Learn More

Linaclotide Ammonium Salt, TRC

Product Code. 30138611
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138611 100 mg 100mg
30009830 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138611 Supplier Toronto Research Chemicals Supplier No. TRCL465885100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C59 H81 N15 O21 S6 . x(H3 N)
Synonym Linzess Ammonium Salt, 11: PN: WO2008151257 SEQID: 55 claimed protein Ammonium Salt 13: PN: WO2008151257 SEQID: 57 claimed protein Ammonium Salt, 18: PN: WO2015054500 SEQID: 72 claimed protein Ammonium Salt, 1: PN: WO2014131024 SEQID: 55 claimed protein Ammonium Salt;1: PN: WO2015054500 SEQID: 55 claimed protein Ammonium Salt;2074: PN: US20090062207 SEQID: 3 claimed protein Ammonium Salt, 3: PN: WO2015054500 SEQID: 57 claimed protein Ammonium Salt, 40: PN: WO2010065751 SEQID: 55 claimed protein Ammonium Salt, 42: PN: WO2010065751 SEQID: 57 claimed protein Ammonium Salt;55: PN: US20100152118 SEQID: 55 claimed protein Ammonium Salt, 55: PN: WO2011069038 SEQID: 55 claimed sequence Ammonium Salt, 55: PN: WO2012037380 SEQID: 55 unclaimed protein Ammonium Salt, 55: PN: WO2015054649 SEQID
SMILES N.C[C@@H](O)C1NC(=O)[C@@H]2CSSC[C@H](NC(=O)[C@@H](N)CSSC[C@H](NC=O)C(=O)N[C@@H](CC(=O)N)C(=O)N3CCC[C@H]3C(=O)N[C@@H](C)C(=O)N2)C(=O)N[C@H](CCC(=O)O)C(=O)N[C@@H](Cc4ccc(O)cc4)C(=O)NCCSSC[C@H](NC(=O)CNC1=O)C(=O)N[C@@H](Cc5ccc(O)cc5)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C59H81N15O21S6.H3N/c1-28-48(83)71-42-26-101-100-25-41(72-49(84)34(60)22-97-99-23-39(64-27-75)52(87)69-37(20-44(61)79)58(93)74-16-3-4-43(74)56(91)65-28)54(89)67-35(13-14-46(81)82)51(86)68-36(18-30-5-9-32(77)10-6-30)50(85)62-15-17-96-98-24-40(66-45(80)21-63-57(92)47(29(2)76)73-55(42)90)53(88)70-38(59(94)95)19-31-7-11-33(78)12-8-31;/h5-12,27-29,34-43,47,76-78H,3-4,13-26,60H2,1-2H3,(H2,61,79)(H,62,85)(H,63,92)(H,64,75)(H,65,91)(H,66,80)(H,67,89)(H,68,86)(H,69,87)(H,70,88)(H,71,83)(H,72,84)(H,73,90)(H,81,82)(H,94,95);1H3/t28-,29+,34-,35+,36-,37-,38-,39-,40-,41-,42-,43-,47?;/m0./s1
Chemical Name or Material Linaclotide Ammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.