Learn More
Memmert™ Light Module Cold White 6,500 K + Warm White 2,700 K
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP110eco
HPP260eco
HPP410eco
HPP750eco
Unit Size:
Each
Description
- 6 resp. 8 alternating cold white light strips and 5 resp
- 7 warm white light strips
- On the side walls of the interior
- Programme-controlled dimming from 0 to 100 % (in 1 % steps)
- Ramp programming in combination with temperature and humidity
- Only withTwinDISPLAY; not in combination with entry ports F6, F7, D8
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | HPP260eco |