Learn More
Invitrogen™ EVOS™ Light Cube, CFP-YFP EM
Click to view available options
Quantity:
Each
Unit Size:
Each
Description
- Contains unique, proprietary LED light cubes
- Image fluorescent samples for specific wavelengthLight engine outputs high intensity over a short light path that delivers superior fluorophore excitation
- Each cube contains an LED, condensing optics, and hard-coated filters
- User-changeable and automatically recognized by system
- Shorter light path that affords superior detection of fluorescent signals
- Continuous illumination that enables consistent results
- 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching
Specifications
Specifications
| Type | Light Cube |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ FL Color Imaging System, Countess™ II FL Automated Cell Counter, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Includes | Condensing Optics, Hard-Coated Filters, LED |
| Wavelength | Ex: 445/45 Em: 542/27 |
| Light Source | Metal Halide (Bulb) |
| Quantity | Each |