Learn More
Invitrogen™ E-Gel™ Imager Camera Hood Replacement Power Cord
Description
Includes
1x E-Gel™ Imager Camera Hood Power Cord
The E-Gel Imager Camera Hood Replacement Power Cord can be used to power the E-Gel Imager Camera Hood. It is a replacement for the power cord that was included in the original package of the E-Gel Imager Camera Hood.
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
| Content And Storage | 1x E-Gel™ Imager Camera Hood Power Cord Store at RT |
| For Use With (Equipment) | E-Gel Imager |
| Product Line | E-Gel |
| Type | Imager Replacement Power Cord |
| Shipping Condition | Room Temperature |
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.