Learn More
Kartell™ Lid for Microtitration Plates
Description
Kartell™ Lid for Microtitration is constructed of polystyrene and can fit plates with flat bottom, U bottom, and V bottom wells.
- For use with multiple-well plates with 96 sample wells
- Manufactured of polystyrene
Specifications
Specifications
| Color | Clear |
| Material | Polystyrene |
| For Use With (Equipment) | Kartell™ 2620, 2621, 2622 |
| Product Type | Lid |