Learn More
Gilac™ Lid with Handle for Round HACCP Food Contact Container
Description
- Lid with handle for round HACCP container.
Specifications
Specifications
| Product Type | HACCP Food Container Lid |
| Material | Polyethylene |
| Color | Blue |
| For Use With (Equipment) | 50 L Round HACCP Food Contact Container |