missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHCGR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19968-1-AP-150UL
This item is not returnable.
View return policy
Description
LHCGR encodes the receptor for both luteinizing hormone and choriogonadotropin. It is family 1 of G-protein coupled receptors, FSH/LSH/TSH superfamily, and its basic function is acting as luteinizing hormone/choriogonadotropin receptor (gonadotropin receptor). Regulating the development and function of Leyding cells in males. It also plays a role in male secondary sexual characterdevelopment. Mutations in this gonadotropinreceptor results in disorders of male secondary sexual characteristic development, such as familial male precocious puperty and male pseudohermaphtoditism.Specifications
| LHCGR | |
| Polyclonal | |
| Unconjugated | |
| LHCGR | |
| LCGR, LGR2, LH/CG R, LH/CGR, LHCGR, LHR, LHRHR, LSH R, Luteinizing Hormone Receptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16867, 25477, 3973 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16235, P22888, P30730 | |
| Lhcgr | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction