missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGR5 Monoclonal Antibody (OTI7F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808752S
This item is not returnable.
View return policy
Description
LGR5 Monoclonal antibody specifically detects LGR5 in Human samples. It is validated for Flow CytometrySpecifications
| LGR5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08183 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5243 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry | |
| OTI7F2 | |
| Unconjugated | |
| ABCB1 | |
| Fex; follicle expressed hormone; G protein-coupled receptor 49; GPR49; GPR67; G-protein coupled receptor 49; G-protein coupled receptor 67; G-protein coupled receptor HG38; GRP49; HG38; leucine rich repeat containing G protein coupled receptor 5; leucine rich repeat containing G protein-coupled receptor 5; leucine-rich repeat containing G protein-coupled receptor 5; leucine-rich repeat-containing G protein-coupled receptor 5; leucine-rich repeat-containing G-protein coupled receptor 5; Lgr5; orphan G protein-coupled receptor HG38; orphan G-protein coupled receptor FEX | |
| Human recombinant protein fragment corresponding to amino acids 995-1280 of human ABCB1 produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction