Learn More
LGALS9 Monoclonal Antibody (OTI21B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805627S
This item is not returnable.
View return policy
Description
LGALS9 Monoclonal antibody specifically detects LGALS9 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| LGALS9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P06400 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5925 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI21B5 | |
| Unconjugated | |
| RB1 | |
| 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; tumor antigen HOM-HD-21; UAT; urate transporter/channel; urate transporter/channel protein | |
| Human recombinant protein fragment corresponding to amino acids 309-590 of human RB1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |