missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGALS9 Monoclonal Antibody (OTI21B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805627
This item is not returnable.
View return policy
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin′s disease tissue and might participate in the interaction between the H&RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
| LGALS9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00182 | |
| LGALS9 | |
| Full length human recombit protein of human LGALS9 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI21B5 | |
| Unconjugated | |
| LGALS9 | |
| HUAT, LGALS9A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3965 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction