Learn More
Leucylisoleucine, TRC
Product Code. 30179183
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179183 100 mg 100mg
30179246 500 mg 500mg
30178958 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179183 Supplier Toronto Research Chemicals Supplier No. TRCA899030100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36077-41-5
Synonym (2S,3S)-2-((S)-2-Amino-4-methylpentanamido)-3-methylpentanoic Acid∣L-Leucyl-L-isoleucine∣165: PN: WO2011146121 PAGE: 117 Claimed Sequence
Specifications
Format Neat
Molecular Formula C12H24N2O3
Molecular Weight (g/mol) 244.33, 244.34
InChI Key AZLASBBHHSLQDB-UHFFFAOYSA-N
SMILES CCC(C)C(NC(=O)C(N)CC(C)C)C(O)=O
IUPAC Name 2-(2-amino-4-methylpentanamido)-3-methylpentanoic acid
Quantity 100 mg
Shipping Condition Room Temperature