missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus, human HPRT

Product Code. 15960353
Click to view available options
Quantity:
100 μL
1 mL
Unit Size:
100µL
1mL
This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15960353

Brand: Invitrogen™ A32056

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.

Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system.

Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice.

The Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice.

Recommended Storage

Store at -68 to -85°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 100 μL. Store (at -68°C to -85°C.)
Control Type Positive Control
Delivery Type Lentiviral
Promoter U6
Transfection Technique Lentiviral transduction
Species Human
For Use With (Application) CRISPR gene knockout
Product Line LentiArray
Product Type CRISPR Positive Control Lentivirus
Quantity 100 μL
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.