Learn More
Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus, human HPRT
Description
Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice.
The Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice.
Specifications
Specifications
| Content And Storage | Store in ultra-cold freezer (-68°C to -85°C). |
| Control Type | Positive Control |
| Delivery Type | Lentiviral |
| Format | 96-well Plate |
| Promoter | U6 |
| Transfection Technique | Lentiviral transduction |
| Species | Human |
| For Use With (Application) | CRISPR gene knockout |
| Product Line | LentiArray |
| Product Type | Positive Control |
| Show More |