Learn More
Cole-Parmer™ Leica Microsystems™ Platinum Calibrated Loops
Shop All Cole Parmer Products
Click to view available options
Volume:
10 μL
Unit Size:
Each
Description
- Ideal for milk cultures.
- Calibrated to American Public Health Association (APHA) standards and made of B&S platinum wire with 5% rhodium
- Insulated loop and needle holder available separately
Specifications
Specifications
| Volume | 10 μL |
| Diameter (Metric) Inside | 4 mm |
| Length (Metric) | 25 mm |
| Material | 19-ga B&S Platinum Wire |
| For Use With (Equipment) | Transferring bacterial cultures |
| Gauge | 19 |
| Calibrated | Yes |
| For Use With (Application) | Transferring bacterial cultures |