missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LEF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582000
This item is not returnable.
View return policy
Description
Immunogen sequence: SDVNSKQGMS RHPPAPDIPT FYPLSPGGVG QITPPLGWQG QPVYPITGGF RQPYPSSLSV DTSMSRFSHH MIPGPPGPHT TGIPHPAIVT PQVKQEHPHT DSDLMHVKPQ HEQRKEQEPK RPHIKKPLNA FMLY Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 96%.
This gene encodes a transcription factor belonging to a family of proteins that share homology with the high mobility group protein-1. The protein encoded by this gene can bind to a functionally important site in the T-cell receptor-alpha enhancer, thereby conferring maximal enhancer activity. This transcription factor is involved in the Wnt signaling pathway, and it may function in hair cell differentiation and follicle morphogenesis. Mutations in this gene have been found in somatic sebaceous tumors. This gene has also been linked to other cancers, including androgen-independent prostate cancer. Alternative splicing results in multiple transcript variants.
Specifications
| LEF1 | |
| Polyclonal | |
| Unconjugated | |
| LEF1 | |
| 3000002B05; AI451430; DKFZp586H0919; FLJ46390; HGNC:6551; HMG Binding Domain; LEF1; Lef-1; lymphoid enhancer binding factor 1; lymphoid enhancer binding factor 1 short isoform; lymphoid enhancer-binding factor 1; OTTHUMP00000219736; OTTHUMP00000219737; T cell-specific transcription factor 1-alpha; TCF10; TCF1ALPHA; TCF1-alpha; TCF7L3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51176 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UJU2 | |
| LEF1 | |
| Recombinant protein corresponding to Human LEF1. Recombinant protein control fragment (Product #RP-100720). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction