Learn More
Lead Standard Solution, 1g/L HNO3 Matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12963195 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 12963195
Brand: Fisher Chemical B 041 500
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Solution Contains HNO3
Concentration 1 g/L
Quantity 500 mL