missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Oil based standard solution, Specpure™ Pb 5000μg/g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| MDL Number | MFCD00134050 |
| Quantity | 50 g |
| Grade | Specpure |
| Chemical Name or Material | Lead, Oil based standard solution, Pb 5000μg/g |
Safety and Handling
Product Identifier
- Lead, Oil based standard solution, Specpure, Pb 5000µg/g
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- Reproductive toxicity on or via lactation
- Reproductive toxicity Category 1A
- H360FD-May damage fertility. May damage the unborn child.
- H362-May cause harm to breast-fed children.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?