Description
- Flexible LDPE construction
- Detachable thread wash head
- Adjustable height delivery tube
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | LDPE |
| No. per Pack | 50/PK |
| Thread Size (Metric) | GL-25 |
| Closure Included | Yes |
| Graduated | No |
| Height (Metric) | 279 mm |
| Product Type | Wash Bottle |
| Diameter (Metric) | 75 mm |