missing translation for 'onlineSavingsMsg'
Learn More

LDN193189, MedChemExpress

Product Code. 16481217
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481217 50 mg 50mg
16457707 5 mg 5mg
16414978 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481217 Supplier Medchem Express Supplier No. HY12071

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LDN193189 (DM-3189) is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM).

LDN193189 (DM-3189) is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM).

Specifications

Chemical Name or Material LDN193189
CAS 1062368-24-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H22N6
Quantity 50 mg
Synonym DM-3189
Solubility Information DMSO : 12 mg/mL (29.52 mM; ultrasonic and adjust pH to 2 with HCl) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C1(C2=C3N=CC(C4=CC=C(N5CCNCC5)C=C4)=CN3N=C2)=CC=NC6=CC=CC=C16
Molecular Weight (g/mol) 406.48
Formula Weight 406.48
Percent Purity 98.9%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.