missing translation for 'onlineSavingsMsg'
Learn More

LDN193189, MedChemExpress

Product Code. 16414978
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16414978

Brand: Medchem Express HY12071

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LDN193189 (DM-3189) is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM).

LDN193189 (DM-3189) is a selective BMP type I receptor inhibitor, which efficiently inhibits ALK2 and ALK3 (IC50=5 nM and 30 nM, respectively), with weaker effects on ALK4, ALK5 and ALK7 (IC50≥500 nM).

Specifications

Chemical Name or Material LDN193189
CAS 1062368-24-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H22N6
Quantity 10 mg
Synonym DM-3189
Solubility Information DMSO : 12 mg/mL (29.52 mM; ultrasonic and adjust pH to 2 with HCl) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C1(C2=C3N=CC(C4=CC=C(N5CCNCC5)C=C4)=CN3N=C2)=CC=NC6=CC=CC=C16
Molecular Weight (g/mol) 406.48
Formula Weight 406.48
Percent Purity 98.9%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.