missing translation for 'onlineSavingsMsg'
Learn More

LDN-214117, MedChemExpress

Product Code. 16403187
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16403187

Brand: Medchem Express HY16712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LDN-214117 is a potent and selective ALK2 inhibitor with IC50 of 22 nM; > 100 fold selectivity for ALK5; also inhibits BMP6(IC50=100 nM).

LDN-214117 is a potent and selective ALK2 inhibitor with IC50 of 22 nM

Specifications

Chemical Name or Material LDN-214117
CAS 1627503-67-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H29N3O3
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (238.37 mM)
SMILES CC(C(C1=CC(OC)=C(OC)C(OC)=C1)=C2)=NC=C2C3=CC=C(N4CCNCC4)C=C3
Molecular Weight (g/mol) 419.52
Formula Weight 419.52
Percent Purity 99.25%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.