missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLR Rabbit anti-Mouse, Clone: 101, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00 EUR - 541.00 EUR
Specifications
| Antigen | LDLR |
|---|---|
| Clone | 101 |
| Dilution | Immunocytochemistry/Immunofluorescence 1:20-1:100 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18288416
|
Novus Biologicals
NBP2-90524-50UL |
50 μg |
367.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18224708
|
Novus Biologicals
NBP2-90524-100UL |
100 μg |
541.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LDLR Monoclonal specifically detects LDLR in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| LDLR | |
| Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
| Recombinant Mouse LDLR Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 101 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 3949 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title