missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3B Antibody (1251D) - BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
470.00 EUR
Specifications
| Antigen | LC3B |
|---|---|
| Clone | 1251D |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 2 ug/ml, Simple Western 5 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Flow (Intracellular) 1 ug/ml, Knockout Validated |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry, KnockDown |
Description
LC3B Monoclonal specifically detects LC3B in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockout Validated.Specifications
| LC3B | |
| 1.0 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| 81631 | |
| Recombinant monoclonal LC3B Antibody (1251D) was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [Uniprot: Q9GZQ8]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1251D | |
| Western Blot 1 - 2 ug/ml, Simple Western 5 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Flow (Intracellular) 1 ug/ml, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| anti-LC3B, ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, LC3B ihc, LC3B immunohistochemistry, LC3B immunoprecipitation, LC3B monoclonal, LC3B western blot, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| MAP1LC3B | |
| IgG | |
| Protein A or G purified | |
| 14.688 kDa |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title