missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lasofoxifene tartrate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Lasofoxifene (CP-336156) tartrate is an orally active and selective estrogen receptor modulator (SERM). Lasofoxifene tartrate exhibits an anti-osteoporotic function and also inhibits primary tumor growth and metastases. Lasofoxifene tartrate can be used for the research of breast cancer and postmenopausal osteoporosis.
Specifications
Specifications
| Chemical Name or Material | Lasofoxifene tartrate |
| CAS | 190791-29-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H37NO8 |
| Quantity | 5 mg |
| Synonym | CP-336156 tartrate |
| Solubility Information | DMSO : 83.33 mg/mL (147.84 mM; Need ultrasonic) |
| SMILES | OC1=CC=C2[C@@H](C3=CC=C(OCCN4CCCC4)C=C3)[C@@H](C5=CC=CC=C5)CCC2=C1.O=C(O)[C@@H](O)[C@H](O)C(O)=O |
| Molecular Weight (g/mol) | 563.64 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction