Learn More
Lasofoxifene tartrate, MedChemExpress
Product Code. 16490708
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16490708
Brand: Medchem Express HYA0038
MedChemExpress Lasofoxifene (CP-336156) tartrate is an orally active and selective estrogen receptor modulator (SERM). Lasofoxifene tartrate exhibits an anti-osteoporotic function and also inhibits primary tumor growth and metastases. Lasofoxifene tartrate can be used for the research of breast cancer and postmenopausal osteoporosis.
Lasofoxifene (CP-336156) tartrate is an orally active and selective estrogen receptor modulator (SERM). Lasofoxifene tartrate exhibits an anti-osteoporotic function and also inhibits primary tumor growth and metastases. Lasofoxifene tartrate can be used for the research of breast cancer and postmenopausal osteoporosis.
Specifications
Chemical Name or Material Lasofoxifene tartrate
CAS 190791-29-8
Color White
Purity Grade Notes Research
Molecular Formula C32H37NO8
Quantity 5 mg
Synonym CP-336156 tartrate
Solubility Information DMSO : 83.33 mg/mL (147.84 mM; Need ultrasonic)
SMILES OC1=CC=C2[C@@H](C3=CC=C(OCCN4CCCC4)C=C3)[C@@H](C5=CC=CC=C5)CCC2=C1.O=C(O)[C@@H](O)[C@H](O)C(O)=O
Molecular Weight (g/mol) 563.64
Formula Weight 563.64
Percent Purity 95.0%
Grade Research
Physical Form Powder