Learn More
Gilac™ Large Volume Container with Pivoting Wheels and Brakes
Product Code. 17908821
Change view
Click to view available options
Capacity (Metric):
100 L
170 L
220 L
310 L
500 L
Includes:
2 Pivoting Wheels with Brakes
4 Pivoting Wheels, 2 Brakes
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes unitSize
17908821 170 L 2 Pivoting Wheels with Brakes Each
17978811 100 L 2 Pivoting Wheels with Brakes Each
17948621 100 L 4 Pivoting Wheels, 2 Brakes Each
17968411 170 L 4 Pivoting Wheels, 2 Brakes Each
17998881 220 L 2 Pivoting Wheels with Brakes Each
17948961 220 L 4 Pivoting Wheels, 2 Brakes Each
17918501 310 L 2 Pivoting Wheels with Brakes Each
17928621 310 L 4 Pivoting Wheels, 2 Brakes Each
17988881 500 L 2 Pivoting Wheels with Brakes Each
17908891 500 L 4 Pivoting Wheels, 2 Brakes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 17908821 Supplier Gilac™ Supplier No. G147131
Only null left
Add to basket
Add to basket
Large Volume Container with Pivoting Wheels and Brakes is the economical solution for moving large volume containers around.
  • The economical solution for moving large volume containers around.
  • Stainless steel wheels and fastenings.
  • Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
  • Load capacity for information purposes only (depending on how the load is spread and the operating temperature).
  • Bottom with gaskets on the axle for perfect sealing.
  • Volume in litres is equivalent to the container filled to the brim.
Specifications
Type Storage Tote
Material HDPE
Color White
Shape Rectangular
Capacity (English) 44.9 gal.
Capacity (Metric) 170 L
Includes 2 Pivoting Wheels with Brakes