Learn More
Cytiva Large Column Holder for O.D. 50mm Columns
Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
Columns with O.D. 50 mm together with AKTAxpress
Unit Size:
Each
Description
Column holder for using columns with o. d. 50mm together with ÅKTA xpress.
Specifications
Specifications
| Description | Large Column Holder |
| For Use With (Equipment) | Columns with O.D. 50 mm together with AKTAxpress |
| Includes | Column holder, Allen key |