missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Laminin alpha-2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579575
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA Cell, A549 Cell, PANC Cell.
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
Specifications
| Laminin alpha-2 | |
| Polyclonal | |
| Unconjugated | |
| Lama2 | |
| 5830440B04; dy; dystrophia muscularis; Lama2; laminin alpha-2; laminin M; Laminin M chain; laminin subunit alpha 2; laminin subunit alpha-2; laminin, alpha 2; laminin-12 subunit alpha; Laminin-2 subunit alpha; laminin-4 subunit alpha; LAMM; mer; merosin; Merosin heavy chain; mKIAA4087 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16773, 309368, 3908 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| P24043, Q60675 | |
| Lama2 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human Laminin 2 alpha (152-170aa WILERSLDDVEYKPWQYHA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction