Learn More
Brady™ Laminated Polyester Stickers for the Transport of Infectious Substances
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Width (Metric):
100.00 mm
200.00 mm
297.00 mm
Unit Size:
Each
Specifications
Specifications
| Alternate Part Number | ADR 6.2-200*200-B7541 |
| Finish | Gloss |
| Material | Laminated Polyester |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| Height (Metric) | 200 mm |
| Thickness | 0.099 mm |
| Width (Metric) | 200.00 mm |
| Temperature Range (Metric) | -40 °C-110 °C |
| Shape | Rhomboid |
| Show More |