missing translation for 'onlineSavingsMsg'
Learn More

Laflunimus Sodium Salt, TRC

Product Code. 30469610
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469610 500 mg 500mg
30480234 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30469610 Supplier Toronto Research Chemicals Supplier No. TRCL168510500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Laflunimus Sodium Salt
Analyte or Component Names Laflunimus Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C15H12F3N2NaO2
Synonym (2Z)-2-Cyano-3-cyclopropyl-3-hydroxy-N-[3-methyl-4-(trifluoromethyl)phenyl]-2-propenamide Sodium Salt, (Z)-2-Cyano-3-cyclopropyl-3-hydroxy-N-[3-methyl-4-(trifluoromethyl)phenyl]-2-propenamide, (Z)-α-Cyano-α4',α4',α4'-trifluoro-β-hydroxycyclopropaneacrylo-3',4'-xylidide Sodium Salt
InChI Formula InChI=1S/C15H13F3N2O2.Na/c1-8-2-5-10(6-12(8)15(16,17)18)20-14(22)11(7-19)13(21)9-3-4-9;/h2,5-6,9,21H,3-4H2,1H3,(H,20,22);/q;+1/p-1/b13-11-;
Solution Type Neat
SMILES O=C(NC1=CC(C(F)(F)F)=C(C)C=C1)/C(C#N)=C(O[Na])/C2CC2
Industry Type Pharmaceutical
Molecular Weight (g/mol) 332.25
Quantity 500 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.