missing translation for 'onlineSavingsMsg'
Learn More

Laduviglusib monohydrochloride, MedChemExpress

Product Code. 16443598
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443598 10 mg 10mg
16429996 50 mg 50mg
16406327 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16443598 Supplier Medchem Express Supplier No. HY10182A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Laduviglusib (CHIR-99021) monohydrochloride is a potent and selective GSK-3α/β inhibitor with IC50s of 10 nM and 6.7 nM.

Laduviglusib monohydrochloride shows >500-fold selectivity for GSK-3 over CDC2, ERK2 and other protein kinases. Laduviglusib monohydrochloride is also a potent Wnt/β-catenin signaling pathway activator. Laduviglusib monohydrochloride enhances mouse and human embryonic stem cells self-renewal. Laduviglusib monohydrochloride induces autophagy.

Specifications

Chemical Name or Material Laduviglusib monohydrochloride
CAS 1797989-42-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H19Cl3N8
Quantity 10 mg
Synonym CHIR-99021 monohydrochlorideCT99021 monohydrochloride
Solubility Information DMSO : 60 mg/mL (119.57 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (14.23 mM; Need ultrasonic)
SMILES N#CC1=CC=C(NCCNC2=NC=C(C3=NC=C(C)N3)C(C4=CC=C(Cl)C=C4Cl)=N2)N=C1.[H]Cl
Molecular Weight (g/mol) 501.8
Formula Weight 501.8
Percent Purity 98.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.