missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lactyl Lysine Monoclonal Antibody (K16286_4E11), Invitrogen™

Product Code. 30201315
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30201315 50 μL 50µg
30198301 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30201315 Supplier Invitrogen™ Supplier No. MA555993

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lactyl lysine is derivative of the amino acid residue lysine where the sidechain ammonium group undergoes lactylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lactyl Lysine
Applications Western Blot
Classification Monoclonal
Clone K16286_4E11
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias l-Lactyl-lysine; Lysine l-lactylation
Host Species Mouse
Immunogen Lactyl Lysine peptide.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.