Learn More
Lacidipine, MedChemExpress
Product Code. p-200098266
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Product Code. 16498448
Brand: Medchem Express HYB0347
MedChemExpress Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.
Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.
Specifications
Chemical Name or Material Lacidipine
CAS 103890-78-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H33NO6
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (109.76 mM)
SMILES O=C(C1=C(C)NC(C)=C(C(OCC)=O)C1C2=CC=CC=C2/C=C/C(OC(C)(C)C)=O)OCC
Molecular Weight (g/mol) 455.54
Formula Weight 455.54
Percent Purity 99.98%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation