Description
- Labworldsoft™ 6 Advanced lab software is unique on the market and the cockpit for your digital laboratory. On the PC, you are able to automate laboratory processes, store your own recipes and process flows or perform calculations during the experiment.
- Labworldsoft™ 6 Advanced at a glance
- Suitable for all device configurations
- Access to more than 700 devices from the device library
- Any number of devices can be controlled at the same time
- For laboratory equipment from all manufacturers
- 1 year of support with new versions of Labworldsoft™ 6 Advanced
- Your benefits
- Time savings and increased safety through automation of processes
- Great flexibility through individual configuration of laboratory processes and set-ups with devices from different manufacturers
- Convenient user interface for a clear display of interlinked work processes
- Easy user management
- Try it free of charge
- You can test Labworldsoft™ 6 Advanced for 30 days free of charge.
- During this time, you have access to the entire device library with over 700 laboratory devices as well as to all functions that are required for your new digital laboratory. There is no obligation for you. After 30 days, you either choose one of our licenses or your access will expire.
Specifications
Specifications
| Type | PC Software |
| For Use With (Application) | Automate Laboratory Processes, Store Recipes and Process Flows or Perform Calculations During The Experiment |