Learn More
Amarell™ Laboratory Total Immersion Precision Thermometer
Specifications
Specifications
| Bottle Fill | Red Spirit |
| Certifications/Compliance | DIN |
| For Use With (Application) | Government testing |
| Immersion Type | Total Immersion |
| Length (Metric) | 220 mm |
| Product Type | Precision Thermometer |
| Max. Temperature (Metric) | 50°C |
| Min. Temperature (Metric) | -10°C |
| Temperature Range (Metric) | -10°C to +50°C |
| Calibrated | Yes |
| Show More |