Learn More
Amarell™ Laboratory Total Immersion Precision Thermometer
Product Code. 10111462
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +100°C
-10°C to +150°C
-10°C to +50°C
Subdivision:
0.1°C
0.5°C
1°C
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Subdivision unitSize
10111462 -10°C to +100°C 0.5°C Each
10726265 -10°C to +150°C 1°C Each
10274672 -10°C to +50°C 0.5°C Each
10084052 -10°C to +50°C 0.1°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10111462 Supplier Amarell™ Supplier No. L26620
Only null left
Specifications
Bottle Fill Red Spirit
Certifications/Compliance DIN
For Use With (Application) Government testing
Immersion Type Total Immersion
Length (Metric) 270 mm
Product Type Precision Thermometer
Max. Temperature (Metric) 100°C
Min. Temperature (Metric) -10°C
Temperature Range (Metric) -10°C to +100°C
Calibrated Yes
Subdivision 0.5°C