Learn More
Amarell™ Laboratory Total Immersion Precision Thermometer
Click to view available options
Temperature Range (Metric):
-10°C to +100°C
-10°C to +150°C
-10°C to +50°C
Subdivision:
0.1°C
0.5°C
1°C
Unit Size:
Each
Specifications
Specifications
| Bottle Fill | Red Spirit |
| Certifications/Compliance | DIN |
| For Use With (Application) | Government testing |
| Immersion Type | Total Immersion |
| Length (Metric) | 420 mm |
| Product Type | Precision Thermometer |
| Max. Temperature (Metric) | 50°C |
| Min. Temperature (Metric) | -10°C |
| Temperature Range (Metric) | -10°C to +50°C |
| Calibrated | Yes |
| Show More |