Learn More
Fisherbrand™ Laboratory Notebooks
Product Code. p-8436003 Shop All Fisherbrand Products
Change view
Click to view available options
Cover Color:
Black
Blue
Green
Red
Ruled:
Grid
Yes
No. of Pages:
100
200
Unit Size:
Pack of 6
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cover Color Ruled No. of Pages unitSize
15811668 Red Yes 200 Pack of 6
15801668 Red Yes 100 Pack of 6
15891658 Red Grid 200 Pack of 6
15881658 Red Grid 100 Pack of 6
15861658 Blue Yes 100 Pack of 6
15851658 Blue Grid 200 Pack of 6
15841658 Blue Grid 100 Pack of 6
15871658 Blue Yes 200 Pack of 6
15831658 Black Yes 200 Pack of 6
15821658 Black Yes 100 Pack of 6
15811658 Black Grid 200 Pack of 6
15801658 Black Grid 100 Pack of 6
16216870 Green Grid 100 Pack of 6
16226870 Green Grid 200 Pack of 6
16236870 Green Yes 100 Pack of 6
16246870 Green Yes 200 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 15811668
Only null left
Add to basket
Add to basket
Keep information safe and well organised with these laboratory notebooks. They are ideal for a variety of applications including recording research data, experimental procedures, design and development ideas.
  • Archival-quality acid-free pages are numbered and include spaces for title, project number, book number, date, and witness signature.
  • Lies flat when opened.
  • User data, notebook instructions, table of contents, periodic table and conversion tables are included in every book.
  • Bookmark ribbon holds your place for quick reference.
  • Covers for all versions are waterproof and chemical-resistant.
  • Lined notebooks feature college-ruled pages for easy report writing.
  • All notebooks contain periodic and conversion tables in the back.
  • Marker ribbon.
  • 8-1/2 x 11 in. (218 x 281 mm)
Note: The line model notebooks have a slight impression of vertical lines. The lines are faint -- only printed at 30%. These vertical lines are helpful if you need to add graphs or drawings to the page.
Specifications
Material Cardboard/Paper
Ruled Yes
Width (English) Page 8.5 in.
Length (English) Page 11 in.
Color Red
Cover Color Red
Dimensions (L x W) 21.8 x 28.1 cm
Length (Metric) Page 27.9 cm
Width (Metric) Page 21.6 cm
Autoclavable Not Autoclavable
Disposable Single-use
No. of Pages 200
Product Type Notebook