Learn More
Epredia™ Laboratory Marking Pen, Secureline Marker II
Shop All Epredia Products
missing translation for 'orderingAttributeHoverText'
Product Type:
Laboratory Marking Pen
Secureline Marker II
missing translation for 'unitSize'
Each
Pack of 10
Description
- Resistant to common laboratory solvents (water, 10% neutral buffered formalin, alcohol)
- Extra fine point for precise writing
- Non-sterile
- Not made with natural rubber latex
Specifications
Specifications
| Product Type | Secureline Marker II |
| Color | Black |